
5-Norbornene-2-ol (endo + exo mixture) 99%
Catalog # H56994.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
Specifications
About this item
CAS Number:13080-90-5
:
Formula:C₇H₁₀O
:110.156 g/mol
Melting Pt:104…106 °C
Flash Pt:51 °C (124 °F)
Storage Temperature:Refrigerator
EINECS:235-987-9
UN:1325
ADR:4.1,III
:GHS02
MDL Number:MFCD00167566