£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United Kingdom
ContactUsLinkComponent
Levofloxacin Q-acid
Levofloxacin Q-acid ≥97%
Levofloxacin Q-acid ≥97%
H54086.03
:
Thermo Fisher Scientific
:
Levofloxacin Q-acid ≥97%
H54086.03
:
Thermo Fisher Scientific
:
Pk:
1 g
:
:
281.21 g/mol
:
>300 °C
:
GHS07
:
Ambient
:
100986-89-8
:
C₁₃H₉F₂NO₄
:
MFCD04039905