sec-Butyl methyl ether 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6795-87-5
:
Formula:C₅H₁₂O
MDL Number:MFCD00190205
:88.15 g/mol
Boiling Pt:57 °C
Density:0.742
Flash Pt:–30 °C (–22 °F)
Storage Temperature:Ambient
ADR:3,II
:GHS02
Frequently Bought Together