Cerium ≥99.9% (REO, rare earth oxide basis), powder < 200 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-45-1
:
Formula:Ce
MDL Number:MFCD00010924
:140.12 g/mol
Merck Index:12,02037
Boiling Pt:2347…2487 °C (1013 hPa)
Melting Pt:815 °C
Density:6.75 g/cm³ (20 °C)
UN:3078
ADR:4.3,II
:Packaged under argon.
EINECS:231-154-9
:GHS02
Storage Temperature:Ambient
Related Information
Frequently Bought Together