£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United Kingdom
ContactUsLinkComponent
3-(Cyanomethyl)benzeneboronic acid
3-(Cyanomethyl)benzeneboronic acid 96%
3-(Cyanomethyl)benzeneboronic acid 96%
H53337.MD
:
Thermo Fisher Scientific
:
3-(Cyanomethyl)benzeneboronic acid 96%
H53337.MD
:
Thermo Fisher Scientific
:
Pk:
250 mg
Seq:
2590
:
220616-39-7
:
:
C₈H₈BNO₂
:
MFCD03788026
:
160.97 g/mol
:
85…88 °C
:
GHS06
:
GHS07
:
Ambient