Order Entry
United Kingdom
ContactUsLinkComponent
4-Diethylcarbamoyl-2-methoxybenzeneboronic acid 96%
Irritant
4-Diethylcarbamoyl-2-methoxybenzeneboronic acid 96%
Catalog # H52837.MD
CAS Number:  
Irritant
4-Diethylcarbamoyl-2-methoxybenzeneboronic acid 96%
Catalog # H52837.MD
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :251.09 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:913835-34-4
Formula:C₁₂H₁₈BNO₄
MDL Number:MFCD09027199