
(S)-1-Boc-2-benzylpiperazine 97%
Catalog # H52785.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:276.38 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:169447-86-3
Formula:C₁₆H₂₄N₂O₂
MDL Number:MFCD03787923