Order Entry
United Kingdom
ContactUsLinkComponent
2-n-Propoxypyridine-3-carboxaldehyde 98%
Irritant
2-n-Propoxypyridine-3-carboxaldehyde 98%
Catalog # H50114.03
CAS Number:  
Irritant
2-n-Propoxypyridine-3-carboxaldehyde 98%
Catalog # H50114.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:885278-12-6
 :
Formula:C₉H₁₁NO₂
MDL Number:MFCD06804561
 :165.19 g/mol
 :GHS07
Storage Temperature:Ambient