Order Entry
United Kingdom
ContactUsLinkComponent
N-(5-Trifluoromethyl-2-pyridyl)thiourea 97%
Toxic
N-(5-Trifluoromethyl-2-pyridyl)thiourea 97%
Catalog # H33731.03
CAS Number:  
Toxic
N-(5-Trifluoromethyl-2-pyridyl)thiourea 97%
Catalog # H33731.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Formula:C₇H₆F₃N₃S
MDL Number:MFCD20921616
 :221.20 g/mol
Melting Pt:202…206 °C
 :GHS06
Storage Temperature:Ambient