
4-Bromo-1-methyl-1H-pyrazole-5-carbaldehyde 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:473528-88-0
:
Formula:C₅H₅BrN₂O
MDL Number:MFCD03074341
:189.02 g/mol
Melting Pt:97…98 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together