Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(4-Bromo-2-nitrophenyl)propionic acid
3-(4-Bromo-2-nitrophenyl)propionic acid 96%
3-(4-Bromo-2-nitrophenyl)propionic acid 96%
H33238.ME
:
Thermo Fisher Scientific
:
3-(4-Bromo-2-nitrophenyl)propionic acid 96%
H33238.ME
:
Thermo Fisher Scientific
:
Pk:
500 mg
Seq:
2725
:
:
274.07 g/mol
:
143…147 °C
:
GHS07
:
Ambient
:
1086386-16-4
:
C₉H₈BrNO₄
:
MFCD11655615