Order Entry
United Kingdom
ContactUsLinkComponent
3-(2,3,3-Trimethyl-3H-indol-5-yl)propionic acid 96%
Irritant
3-(2,3,3-Trimethyl-3H-indol-5-yl)propionic acid 96%
Catalog # H33049.03
CAS Number:  
Irritant
3-(2,3,3-Trimethyl-3H-indol-5-yl)propionic acid 96%
Catalog # H33049.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :231.29 g/mol
Melting Pt:145…146 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:1072069-91-0
Formula:C₁₄H₁₇NO₂
MDL Number:MFCD12922972