Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(3-Bromo-2-fluorophenyl)acetonitrile
(3-Bromo-2-fluorophenyl)acetonitrile 98%
(3-Bromo-2-fluorophenyl)acetonitrile 98%
H32021.MD
:
Thermo Fisher Scientific
:
(3-Bromo-2-fluorophenyl)acetonitrile 98%
H32021.MD
:
Thermo Fisher Scientific
:
Pk:
250 mg
Seq:
2590
:
:
214.04 g/mol
:
92 °C/0.3 mm
:
3276
:
6.1,III
:
GHS06
:
874285-03-7
:
C₈H₅BrFN
:
MFCD09864662