Order Entry
United Kingdom
ContactUsLinkComponent
 
3,3,3-Trifluoropropane-1-sulphonyl chloride 95%
Catalog # H25856.ME
CAS Number:  
3,3,3-Trifluoropropane-1-sulphonyl chloride 95%
Catalog # H25856.ME
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 3,3,3-Trifluoropropane-1-sulphonylchloride
CAS Number:845866-80-0
 :
Formula:C₃H₄ClF₃O₂S
MDL Number:MFCD06248898
 :196.58 g/mol
 :GHS05
Boiling Pt:77…78 °C (16 mmHg)
Storage Temperature:Ambient
ADR:8 II