2-Ethylthio-1,3,4-thiadiazole-5-thiol
Catalog # B22464.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:178.298 g/mol
CAS Number:37147-15-2
Formula:C₄H₆N₂S₃
MDL Number:MFCD00186131