N-Boc-L-alanine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15761-38-3
:
Formula:C₈H₁₅NO₄
MDL Number:MFCD00037225
:189.21 g/mol
Melting Pt:80…84 °C
EINECS:239-847-8
Related Information
Frequently Bought Together