Rhodium 5% on carbon, dry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-16-6
:
Formula:Rh
MDL Number:MFCD00011201
Merck Index:14,8186
ADR:4.1,III
:102.9 g/mol
EINECS:231-125-0
UN:1325
:GHS02
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together