Diethyl diallylmalonate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3195-24-2
:
Formula:C₁₃H₂₀O₄
MDL Number:MFCD00009126
:240.30 g/mol
Boiling Pt:242…244 °C
Density:0.993
Flash Pt:>110 °C (230 °F)
EINECS:221-696-4
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together