Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Antimony
Antimony ≥99.999% (metals basis), rods, Ø 12.7 mm (0.500 in)
Antimony ≥99.999% (metals basis), rods, Ø 12.7 mm (0.500 in)
47181.13
:
Thermo Fisher Scientific
:
Antimony ≥99.999% (metals basis), rods, Ø 12.7 mm (0.500 in)
47181.13
:
Thermo Fisher Scientific
:
Pk:
20 g
Seq:
3290
:
:
121.76 g/mol
:
Ambient
:
231-146-5
:
2871
:
6.1,III
:
7440-36-0
:
Sb
:
MFCD00134030