

Chromium etchant
Catalog # 44582.WY
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:950 mL
- Seq:2845
Specifications
About this item
:
Density:1.16
Storage Temperature:Ambient
UN:3093
ADR:8,II
:GHS03
:GHS05
MDL Number:MFCD07370678