Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Nickel plating solution, electroless for copper and copper alloys

1 L
Catalog # 44215.K2
Health
Irritant
Nickel plating solution, electroless for copper and copper alloys
Catalog # 44215.K2
CAS Number:  
Nickel plating solution, electroless for copper and copper alloys
Nickel plating solution, electroless for copper and copper alloys

1 L

CAS Number:  
Price
£74.20
Log in for availability
1 * 1 L
Product Details & Documents

Appearance: Clear green UN No.: UN3082 MDL No.: MFCD00011137

Applications: For plating on copper, brass and copper alloys

  • MDL Number:MFCD00011137
  • Storage Temperature:Ambient
  • UN:3082
  • ADR:9,III
Specifications

Specifications

  • Pk:
    1 L

Specifications