
Bis[3-(triethoxysilyl)propyl]tetrasulphide (∼22.3% S) ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:40372-72-3
:
Formula:C₁₈H₄₂O₆S₄Si₂
MDL Number:MFCD00053751
:538.938 g/mol
Boiling Pt:250 °C (dec.)
Density:1.074
Flash Pt:91 °C (196 °F)
EINECS:254-896-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together