Erbium(III)-2,4-pentanedionate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14553-08-3
:
Formula:C₁₅H₂₁ErO₆
MDL Number:MFCD00013490
:464.59 g/mol
Storage Temperature:Ambient
Related Information
Frequently Bought Together