Rhodium 5% on alumina, powder, reduced
Catalog # 11770.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:10 g
Specifications
About this item
:
:102.9 g/mol
Storage Temperature:Ambient
EINECS:231-125-0
CAS Number:7440-16-6
Formula:Rh
MDL Number:MFCD00011201