Rhodium 1% on alumina, powder, reduced
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-16-6
:
Formula:Rh
MDL Number:MFCD00011201
:102.9 g/mol
EINECS:231-125-0
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together