Order Entry
United Kingdom
ContactUsLinkComponent
Horizontal electrophoresis system, multiSUB maxi
Horizontal electrophoresis system, multiSUB maxi
Supplier:  CLEAVER SCIENTIFIC
Certificates
undefined
Horizontal electrophoresis system, multiSUB maxi
Supplier:  CLEAVER SCIENTIFIC
Specifications
  • Catalog No:
  • CLEVMSMAXI25
  • CLEVMSMAXI20
  • CLEVMSMAXIDUO
  • Model:
  • multiSUB maxi 25
  • multiSUB maxi 20
  • multiSUB maxi DUO
  • Gel size W×L:
  • 200×250 mm
  • 200×200 mm
  • 200×100 mm; 200×200 mm
  • Sample capacity:
  • 550 samples
  • 450 samples
  • Buffer volume:
  • 1200 ml
  • 1200 ml
  • 1200 ml
  • Unit W×L×H:
  • 230×395×90 mm
  • 230×395×90 mm
  • 230×395×90 mm
Specifications
Combs
Product Information
Frequently Bought Together