Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Horizontal electrophoresis system, multiSUB mini
Horizontal electrophoresis system, multiSUB mini
Supplier:   CLEAVER SCIENTIFIC
Horizontal electrophoresis system, multiSUB mini
Supplier:  CLEAVER SCIENTIFIC
Certificates
Horizontal electrophoresis system, multiSUB mini
3 Options Available Below
Product Details & Documents
The multiSUB mini is designed for low to medium numbers of samples. The small gel size maximises run economy but does not compromise versatility as two tray options are available – 70×70 mm and 70×100 mm – and combs ranging from preparative up to 16 samples. Simply by altering the gel tray or comb, this compact unit is capable of resolving up to 64 different samples, prepping 1 ml of sample or separating sample bands over a distance of 9 cm.
  • Two tray options
  • Run up to 64 samples
  • Low buffer volumes
Specifications for Product Family
Specifications
  • Catalog No:
  • CLEVMSMINI7
  • CLEVMSMINI10
  • CLEVMSMINIDUO
  • Model:
  • multiSUB mini 7
  • multiSUB mini 10
  • multiSUB mini DUO
  • Gel size W×L:
  • 70×70 mm
  • 70×100 mm
  • 70×100 mm; 70×100 mm
  • Sample capacity:
  • 32 samples
  • 64 samples
  • Buffer volume:
  • 225 ml
  • 225 ml
  • 225 ml
  • Unit W×L×H:
  • 90×210×90 mm
  • 90×210×90 mm
  • 90×210×90 mm
Specifications
Product Family Options
Product Information
  • Availability
    Price
  • CLEVMSMINI7
    multiSUB mini 7, 70×70 mm, 2× 8-sample combs, loading guides and dams
    multiSUB mini 7, 70×70 mm, 2× 8-sample combs, loading guides and dams
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - £531.00
    £531.00
    1 * 1 items
       £531.00
    Specifications:
    More
    Specifications
    Cat. No.
    CLEVMSMINI7
    Model
    multiSUB mini 7
    Gel size W×L
    70×70 mm
    Sample capacity
    32 samples
    Buffer volume
    225 ml
    Unit W×L×H
    90×210×90 mm
  • CLEVMSMINI10
    multiSUB mini 10, 70×100 mm, 2× 8-sample combs, loading guides and dams
    multiSUB mini 10, 70×100 mm, 2× 8-sample combs, loading guides and dams
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - £517.00
    £517.00
    1 * 1 items
       £517.00
    Specifications:
    More
    Specifications
    Cat. No.
    CLEVMSMINI10
    Model
    multiSUB mini 10
    Gel size W×L
    70×100 mm
    Sample capacity
    64 samples
    Buffer volume
    225 ml
    Unit W×L×H
    90×210×90 mm
  • CLEVMSMINIDUO
    multiSUB mini DUO, 70×70 mm and 70×100 mm, 2× 8-sample combs, loading guides and dams
    multiSUB mini DUO, 70×70 mm and 70×100 mm, 2× 8-sample combs, loading guides and dams
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - £561.00
    £561.00
    1 * 1 items
       £561.00
    Specifications:
    More
    Specifications
    Cat. No.
    CLEVMSMINIDUO
    Model
    multiSUB mini DUO
    Gel size W×L
    70×100 mm; 70×100 mm
    Sample capacity
    Buffer volume
    225 ml
    Unit W×L×H
    90×210×90 mm
Combs
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Comb, 16-sample, 1,5 mm thick
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - £49.60
    £49.60
    1 * 1 items
       £49.60
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Comb, 16-sample, 1 mm thick
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - £49.60
    £49.60
    1 * 1 items
       £49.60
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Comb, 8-sample, 0,75 mm thick*
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - £0.00
    £0.00
    1 * 1 items
       £0.00
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Comb, 16-sample, 0,75 mm thick
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - £51.00
    £51.00
    1 * 1 items
       £51.00
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Negative electrode
    Negative electrode
    CLEVMS7-NE
    Log in for availability
    1 * 1 items - £135.00
Product Information
  • Catalog #
    Availability
    Price
  • Positive electrode
    Positive electrode
    CLEVMS7-PE
    Log in for availability
    1 * 1 items - £135.00
Product Information
  • Catalog #
    Availability
    Price
  • UV tray, 70×70 mm
    UV tray, 70×70 mm
    CLEVMS7-UV7
    Log in for availability
    1 * 1 items - £83.90