Calcium phosphate tribasic, powder NF, FCC, Macron Fine Chemicals™
:1306-06-5
:12167-74-7
:Ca₅HO₁₃P₃
:MFCD00010904
:502.306 g/mol
:Management of Change (MOC) category = R
- Meets F.C.C. Requirements:—
- GMP Manufactured Product:—
- Meets N.F. Requirements:—
- Food GMP Manufactured Product:—
- CAUTION: For Manufacturing, processing or repackaging:—
- Bulk Food Chemical:—
- Bulk Pharmaceutical Chemical:34.0 - 40.0 %
- NF - Assay (Ca):34.0 - 40.0 %
- NF - Acid-Insoluble Substances:≤ 0.2 %
- NF - Arsenic (As):≤ 3 mg/kg
- NF - Barium (Ba):Passes Test
- NF - Carbonate (CO₃):Passes Test
- NF - Chloride (Cl):≤ 0.14 %
- NF - Dibasic Salt and Calcium Oxide (ml):13.0 - 14.3
- NF - Fluoride (F):≤ 0.0075 %
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Loss on Ignition:≤ 8.0 %
- NF - Nitrate (NO₃):Passes Test
- NF - Sulfate (SO₄):≤ 0.8 %
- NF - Water-Soluble Substances:≤ 0.5 %
- FCC - Assay (Ca):34.0 - 40.0 %
- FCC - Arsenic (As):≤ 3 mg/kg
- FCC - Fluoride (F):≤ 0.0075 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Lead (Pb):≤ 2 mg/kg
- FCC - Loss on Ignition:≤ 8.0 %
Frequently Bought Together