Burette clamp, double, Talon®
Catalog # TROE915028
Supplier: TROEMNER INSTRUMENTS
Specifications
- For:Two burettes (max. 100 ml)
- Description:Double burette clamp
- Pk:10
Specifications
About this item
This research grade clamp holds two burettes from micro to 100 ml size.
- Scissors-like mechanism adjusts simply with a squeeze, yet grips firmly
- Clamp unit attaches to a standard support rod with a built-in reinforced hook connector
Ordering information: The support rod is available separately. Please contact your local Avantor sales office for further information.