Surface cleansing wipes, dirteeze rough & smooth, heavy duty
Catalog # ESPADGPCL80
Supplier: ECOSPILL LIMITED
Specifications
- Packed:80 wipes/tub dispenser
- L×W:220×200 mm
- Pk:8
Specifications
About this item
These pre-moistened wipes are ideal for cleaning rough surfaces, machinery and tools.
- Removes oils, greases, semi-cured sealants and adhesives
- Removes water-based paints and general grime
- Safe for hands, tools and surfaces
- Contain aloe vera to keep hands moisturised
- Anti-bacterial additive kills most known germs
- Great in engineering, construction and industrial sectors
The wipes are dual-sided with a micro-beaded surface on one side, which can be used to break up, agitate and remove any stubborn grime. Then simply use the smooth side to wipe away.