Order Entry
United Kingdom
ContactUsLinkComponent
Craft blades, Slice®
Catalog # KNIFSLICECRAFTBLAD
Supplier:  THE SAFETY KNIFE
undefined
Craft blades, Slice®
Catalog # KNIFSLICECRAFTBLAD
Supplier:  THE SAFETY KNIFE
Specifications
  • Description:
    Craft blades for Slice® Craft Knife, Precision Knife, Scalpel
  • L×W:
    32,8×6,3 mm
  • Material:
    Zirconium oxide
  • Thickness:
    0,5 mm
  • Pk:
    1
Specifications
About this item
These Slice® craft blades provide controlled, detailed cuts and a much safer finger-friendly® cutting edge.
  • Lasts up to 10× longer than steel blades
  • Straight edge with safer rounded tip
  • Does not conduct electricity
  • Never rusts, chemically inert, non-magnetic
Slice® craft blades are designed to work exclusively with Slice® handles for optimal safety and function.