Carbon (from wood), powder, Millipore®
Catalog # 1.02204.9025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Fibre carton
- Pk:25 kg
- Seq:4283
Specifications
Specification Test Results
- Residue of ignition (600 °C):≤7%
- Particle size <0.15 mm:~99%
- Carbonization:Conforms
- Bulk Density:410 kg/m3
- Hill Formula:C
- Chemical Formula:C
- Melting point:3550 °C
- Residue on ignition :≤7 %
- Particle size:about 99 %
Specification Test Results
About this item
:
:12.01 g/mol
ADR:4.2,III
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:231-153-3
UN:1361
CAS Number:7440-44-0
Formula:C
MDL Number:MFCD00133992