Anti-CTGF Rabbit Polyclonal Antibody
Catalog # PIEAPA532193
Supplier: Thermo Fisher Scientific
Specifications
- Antibody type:Primary
- Antigen name:Connective Tissue Growth Factor
- Antigen symbol:CTGF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:64032, 14219, 1490
- Antigen synonyms:IGFBP8|hypertrophic chondrocyte-specific protein 24|HCS24|IGFBP-8|insulin-like growth factor-binding protein 8|IBP-8|IGF-binding protein 8|CCN2|CCN family member 2|NOV2
- Storage buffer:PBS, 1% BSA, 20% glycerol, 0.01% thimerosal, pH 7
- Storage temperature:-20° C, Avoid Freeze/Thaw Cycles
- Concentration:1 mg/ml
- Immunogen:Recombinant fragment corresponding to a region within amino acids 115 and 315 of Human CTGF
- Tested applications:Immunohistochemistry (Paraffin)
- Purification:Antigen affinity chromatography
- Size:100 µL
- Pk:100 µl
Specifications
About this item
PA5-32193 targets CTGF in IHC (P) and WB applications and shows reactivity with Human, mouse, and Rat samples. The PA5-32193 immunogen is recombinant fragment corresponding to a region within amino acids 115 and 315 of Human CTGF. The protein encoded by this gene is a mitogen that is secreted by vascular endothelial cells. The encoded protein plays a role in chondrocyte proliferation and differentiation, cell adhesion in many cell types, and is related to platelet-derived growth factor. Certain polymorphisms in this gene have been linked with a higher incidence of systemic sclerosis.
- Immunohistochemistry (Paraffin): 1:100-1:1000, Western Blot: 1:1000-1:10,000
Type: Primary
Antigen: CTGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat