Anti-ZNRF3 Rabbit Polyclonal Antibody
Catalog # BOSSBS-9141R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Zinc And Ring Finger 3
- Antigen symbol:ZNRF3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Form:liquid
- Gene ID:84133
- Antigen synonyms:BK747E2.3|E3 ubiquitin-protein ligase ZNRF3|Zinc/RING finger protein 3|RNF203|KIAA1133|RING finger protein 203|ZNRF3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Immunogen:120-165/936
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
E3 ubiquitin-protein ligase that acts as a negative regulator of the Wnt signaling pathway by mediating the ubiquitination and subsequent degradation of Wnt receptor complex components Frizzled and LRP6. Acts on both canonical and non-canonical Wnt signaling pathway. Acts as a tumor suppressor in the intestinal stem cell zone by inhibiting the Wnt signaling pathway, thereby resticting the size of the intestinal stem cell zone.
IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: ZNRF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat