Anti-HMGCS1 Rabbit Polyclonal Antibody
BOSSBS-5069R
: Bioss
- Antibody type:Primary
- Antigen name:3-hydroxy-3-methylglutaryl-CoA Synthase 1 (soluble)
- Antigen symbol:HMGCS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:266977
- Antigen synonyms:HMGCS|3 hydroxy 3 methylglutaryl Coenzyme A synthase 1 soluble|cytoplasmic|MGC90332: EC 2.3.3.10|HMCS1_HUMAN.|Hydroxymethylglutaryl CoA synthase|HMG CoA synthase
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
This enzyme condenses acetyl CoA with acetoacetyl CoA to form HMG CoA, which is the substrate for HMG CoA reductase.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: HMGCS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat