Anti-PHLDA1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-1364R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Pleckstrin homology-like domain A1
- Antigen symbol:PHLDA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:22822
- Antigen synonyms:PHRIP|T-cell death-associated gene 51 protein|Proline- and glutamine-rich protein|PHLDA1|Pleckstrin homology-like domain family A member 1|Proline- and histidine-rich protein|PQ-rich protein|PQR protein|TDAG51|Apoptosis-associated nuclear protein
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Immunogen:265-305/401
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Seems to be involved in regulation of apoptosis. May be involved in detachment-mediated programmed cell death. May mediate apoptosis during neuronal development. May be involved in regulation of anti-apoptotic effects of IGF1. May be involved in translational regulation.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: PHLDA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat