Anti-RBC Rabbit Monoclonal Antibody (Unconjugated) [clone: BM3]
Catalog # ROCK112-4139
Supplier: Rockland Immunochemicals
Specifications
- Antigen name:Rat washed pooled Red Blood Cells
- Antigen symbol:RBC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Rat
- Format:Antiserum
- Form:Lyophilized
- Storage buffer:Restore with deionized water (or equivalent), 2.0 ml
- Concentration:90 mg/ml by Refractometry
- Shipping temperature:Ambient
- Immunogen:Rat washed pooled Red Blood Cells (RBC)
- Purification:This product was prepared from polyspecific antiserum by a delipidation and defibrination.
- Size:2 ml
- Pk:2 mL
Specifications
About this item
contains 0.01% (w/v) Sodium Azide
Type: Primary
Antigen: RBC
Clonality: Monoclonal
Clone: BM3
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat