Order Entry
United Kingdom
ContactUsLinkComponent
Dual plate rotators
Dual plate rotators
Catalog # HARV74-2334
Supplier:  HARVARD APPARATUS
undefined
Dual plate rotators
Catalog # HARV74-2334
Supplier:  HARVARD APPARATUS
Specifications
  • Description:
    Dual plate rotator
  • Pk:
    1
Specifications
About this item
Dual plate rotator with variable rotation rates, can be used with the Harvard apparatus 96-well equilibrium dialyser. The rotator speeds up the equilibrium dialysis process by keeping the sample in constant motion thereby ensuring higher reproducibility of results.