VWR® Bottles, Wide Neck, with Thread
Catalog # VWRL0340P00042
Supplier: LAB3 OWN PRODUCTION
Specifications
- Thread:GL 55
- Capacity:250 ml
- Pk:1
Specifications
About this item
Clear glass with thread.
- Good chemical and physical properties
- For applications that do not involve high thermal stresses
Ideal for sampling, storing and transporting powders and pastes. Screw cap must be ordered separately.