Anti-GFAP Antibody Pair
Catalog # ABNOH00002670-AP52
Supplier: Abnova
Specifications
- Antigen name:Glial fibrillary acidic protein
- Antigen symbol:GFAP
- Host:Multiple
- Reactivity:Human
- Gene ID:2670
- Antigen synonyms:FLJ45472|Intermediate filament protein.|Astrocyte|p-GFAP Ser38|GFAP phospho S38|GFAP|Glial Fibrillary Acidic Protein
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
Specifications
About this item
This Set comes with matched Antibody Pair to detect and quantify protein level of human GFAP.
Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-GFAP, IgG2a kappa (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-GFAP (50 µg);
Type:
Antigen: GFAP
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human