Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-SMPDL3B Polyclonal Antibody Pair
  ABNOH00027293-AP21
 :  Abnova
Anti-SMPDL3B Polyclonal Antibody Pair
  ABNOH00027293-AP21
 :  Abnova

 

  • Antigen name:
    Sphingomyelin Phosphodiesterase, Acid-like 3B
  • Clonality:
    Polyclonal
  • Host:
    Multiple
  • Reactivity:
    Human
  • Gene ID:
    27293
  • Antigen synonyms:
    ASML3B
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

 

 

This Set comes with matched Antibody Pair to detect and quantify protein level of human SMPDL3B.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-SMPDL3B (100 µg); 2. Detection Antibody: Mouse Polyclonal Anti-SMPDL3B (40 µl);

Type:
Antigen: SMPDL3B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human