Order Entry
United Kingdom
ContactUsLinkComponent
Anti-PSAP Antibody Pair
Anti-PSAP Antibody Pair
Catalog # ABNOH00005660-AP11
Supplier:  Abnova
Anti-PSAP Antibody Pair
Catalog # ABNOH00005660-AP11
Supplier:  Abnova

Specifications

  • Antigen name:
    Prostate Specific Acid Phosphatase
  • Antigen symbol:
    PSAP
  • Host:
    Multiple
  • Reactivity:
    Human
  • Gene ID:
    5660
  • Antigen synonyms:
    SAP1|PAP|prostatic acid phosphotase|prostatic acid phosphatase|GLBA
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

Specifications

About this item

This Set comes with matched Antibody Pair to detect and quantify protein level of human PSAP.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-PSAP (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-PSAP, IgG2a kappa (20 µg);

Type:
Antigen: PSAP
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human