Anti-FABP1 Antibody Pair
Catalog # ABNOH00002168-AP13
Supplier: Abnova
Specifications
- Antigen name:Fatty Acid Binding Protein 1
- Antigen symbol:FABP1
- Host:Multiple
- Reactivity:Human
- Gene ID:2168
- Antigen synonyms:SCP|FABPL|p14|SCP.|L-FABP
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
Specifications
About this item
This Set comes with matched Antibody Pair to detect and quantify protein level of human FABP1.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-FABP1 (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-FABP1, IgG2b kappa (20 µg);
Type:
Antigen: FABP1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human