Order Entry
United Kingdom
ContactUsLinkComponent
Anti-SIGLEC6 Antibody Pair
Anti-SIGLEC6 Antibody Pair
Catalog # ABNOH00000946-AP51
Supplier:  Abnova
Anti-SIGLEC6 Antibody Pair
Catalog # ABNOH00000946-AP51
Supplier:  Abnova

Specifications

  • Antigen name:
    Sialic acid binding Ig-like lectin 6
  • Antigen symbol:
    SIGLEC6
  • Host:
    Multiple
  • Reactivity:
    Human
  • Gene ID:
    946
  • Antigen synonyms:
    CD33L|CD327|Sialic acid binding Ig-like lectin 6|Obesity-binding protein 1|OBBP1|CD33L2|CD33 antigen-like 1|CDW327|CD33L1|OB-BP1
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

Specifications

About this item

This Set comes with matched Antibody Pair to detect and quantify protein level of human SIGLEC6.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-SIGLEC6 (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-SIGLEC6 (50 µg);

Type:
Antigen: SIGLEC6
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human