Order Entry
United Kingdom
ContactUsLinkComponent
Anti-TFB1M Antibody Pair
Anti-TFB1M Antibody Pair
  ABNOH00051106-AP51
 :  Abnova
Anti-TFB1M Antibody Pair
  ABNOH00051106-AP51
 :  Abnova

 

  • Antigen name:
    Transcription factor B1, mitochondrial
  • Antigen symbol:
    TFB1M
  • Host:
    Multiple
  • Reactivity:
    Human
  • Gene ID:
    51106
  • Antigen synonyms:
    homolog of yeast mitochondrial transcription factor B|N'-adenosyl(rRNA) dimethyltransferase 1|mitochondrial 12S rRNA dimethylase 1|S-adenosylmethionine-6-N'|mtTFB|CGI-75|mitochondrial|CGI75|transcription factor B1|OTTHUMP00000017473|mtTFB1|TFB1M
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

 

 

This Set comes with matched Antibody Pair to detect and quantify protein level of human TFB1M.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-TFB1M (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-TFB1M (50 µg);

Type:
Antigen: TFB1M
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human