Anti-FABP1 Polyclonal Antibody Pair
ABNOH00002168-AP21
: Abnova
- Antigen name:Fatty Acid Binding Protein 1
- Antigen symbol:FABP1
- Clonality:Polyclonal
- Host:Multiple
- Reactivity:Human
- Gene ID:2168
- Antigen synonyms:SCP|FABPL|p14|SCP.|L-FABP
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
This Set comes with matched Antibody Pair to detect and quantify protein level of human FABP1.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-FABP1 (100 µg); 2. Detection Antibody: Mouse Polyclonal Anti-FABP1 (40 µl);
Type:
Antigen: FABP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human