Order Entry
United Kingdom
ContactUsLinkComponent
Anion exchange columns, Dionex™, IonPac™ AS19
 :  DIONEX
Anion exchange columns, Dionex™, IonPac™ AS19
 :  DIONEX
  • Catalog No:
  • DION062887
  • DION062886
  • DION062885
  • DION062888
  • Max. pressure:
  • 3000 psi
  • 3000 psi
  • 3000 psi
  • 3000 psi
  • pH range:
  • 0 - 14
  • 0 - 14
  • 0 - 14
  • 0 - 14
Frequently Bought Together