QC organisms and sets, LyfoCults® Plus
BMER301072
:
- Pk:1
- Description:LyfoCults Plus Enterobacter hormaechei ATCC® 700323
LyfoCults Plus is a storage, hydration and inoculation system. A technologically advanced microorganism system on the market. Other systems use glass in the device. Convenient QC Sets are also available for all identification products (VITEK®2 Compact, API®/ID32). Available in two packs.
- The patented 'All-In-One' design is simple and easy to use
- Built in inoculation wand that eliminates the need for additional tools and reduces contamination
- Glass free to improve safety
- Hydrate immediately saving time