Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-CTBP1 Rabbit Polyclonal Antibody
  ABNOPAB29240
 :  Abnova
Anti-CTBP1 Rabbit Polyclonal Antibody
  ABNOPAB29240
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    C-terminal binding protein 1
  • Antigen symbol:
    CTBP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    1487
  • Antigen synonyms:
    BARS
  • Modification:
    phospho-Ser422
  • Storage buffer:
    PBS, pH 7,4 (without Mg(2+) and Ca(2+)), (50% glycerol, 0,02% sodium azide)
  • Storage temperature:
    Store at –20 °C. Avoid repeated freezing and thawing.
  • Immunogen:
    Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S422 of human CTBP1.
  • Purification:
    Affinity purification
  • Size:
    100 μl
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody raised against synthetic phosphopeptide of human CTBP1.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100; Western Blot: 1:500-1000

Type: Primary
Antigen: CTBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: