Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-FAS Mouse Monoclonal Antibody [clone: 5B4]
  ABNOH00000355-M01A
 :  Abnova
Anti-FAS Mouse Monoclonal Antibody [clone: 5B4]
  ABNOH00000355-M01A
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Fatty Acid Synthase
  • Antigen symbol:
    FAS
  • Clonality:
    Monoclonal
  • Clone:
    5B4
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Form:
    Ascites fluid
  • Gene ID:
    355
  • Antigen synonyms:
    FASTM|APO-1|ALPS1A|TNFRSF6|CD95|APT1|FAS1
  • Amino acid number:
    20 to 119
  • Sequence:
    SKSVNAQVTDINSKGLELRKTVTTVETQNLEGLHHDGQFCHKPCPPGERKARDCTVNGDEPDCVPCQEGKEYTDKAHFSSKCRRCRLCDEGHGLEVEINC
  • Storage temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    FAS (NP_000034, 20 a.a. ~ 119 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
  • Size:
    200 μl
  • Pk:
    200 µg

 

 

Mouse monoclonal antibody raised against a partial recombinant FAS.

Type: Primary
Antigen: FAS
Clonality: Monoclonal
Clone: 5B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: